Learn More
Uvex Fold-flat Mask silv-Air Premium 5220+ FFP2
Product Code. 17572975 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17572975 Filter Type: FFP2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17572975 Supplier Uvex Supplier No. 8765221
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The silv-Air premium+ product line comprises classic folding respirators that adapt to different face shapes which fits perfectly to larger face shapes. The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
General Features
  • Individually packed
  • Color: gray
  • With exhalation valve
Safety Features
  • Certified FFP2 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Additional activated carbon filter against odours, gases and vapours below threshold
  • Cannot be reused
Comfort Features
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
  • Adjustable head band offering easy handling and comfortable fit
  • Flexible and adjustable nose clip
  • Soft material edges for high wearing comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Disposable Single-use
Material Polypropylene, Polyester
With Exhalation Valve Yes
Filter Efficiency FFP2
Sterility Non-sterile