missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid, ≥95%

Product Code. 11435162 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435162 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11435162 Supplier MP Biomedicals Supplier No. 0219466525
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 59-30-3
Color Yellow to Orange
Molecular Formula C19H19N7O6
Quantity 25 g
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 441.404
PubChem CID 6037
ChEBI CHEBI:27470
Formula Weight 441.4
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Folic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.