missing translation for 'onlineSavingsMsg'
Learn More

Foretinib, TRC

Product Code. 30402762
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402762 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402762 Supplier Toronto Research Chemicals Supplier No. TRCF691200100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Foretinib
CAS 849217-64-7
Molecular Formula C34H34F2N4O6
Quantity 100 mg
Synonym EXEL 2880, GSK 1363089, GSK 1363089G, N-[3-Fluoro-4-[[6-(methyloxy)-7-[[3-(morpholin-4-yl)propyl]oxy]quinolin-4-yl]oxy]phenyl]-N'-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL 880
InChI Formula InChI=1S/C34H34F2N4O6/c1-43-30-20-25-27(21-31(30)45-16-2-13-40-14-17-44-18-15-40)37-12-9-28(25)46-29-8-7-24(19-26(29)36)39-33(42)34(10-11-34)32(41)38-23-5-3-22(35)4-6-23/h3-9,12,19-21H,2,10-11,13-18H2,1H3,(H,38,41)(H,39,42)
SMILES COc1cc2c(Oc3ccc(NC(=O)C4(CC4)C(=O)Nc5ccc(F)cc5)cc3F)ccnc2cc1OCCCN6CCOCC6
Molecular Weight (g/mol) 632.65
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.