missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Standard Solution 1000 ppm, Chem Lab

Product Code. 11949062
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11949062 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11949062 Supplier Chem-Lab Supplier No. CL01.0651.0100
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Polyethylene
CAS 50-00-0
Solution Contains 1000 μg/mL of HCHO
Density 1 g/mL
Physical Form Liquid
Concentration 1000 μg/mL
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Weight (g/mol) 30.03
Quantity 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.