Learn More
Melford Formalin
Product Code. 30018272
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018272 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018272 Supplier Melford Supplier No. F10800500.0
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Widely used tissue fixative for common histology applications.
Widely used tissue fixative for common histology applications. Ready to use formula does not require dilution.
Specifications
Quantity 500 mL