missing translation for 'onlineSavingsMsg'
Learn More

Formestane, TRC

Product Code. 30134135
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134135 1 g 1g
30007611 50 mg 50mg
30007954 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134135 Supplier Toronto Research Chemicals Supplier No. TRCF6922501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-48-3
Molecular Formula C19 H26 O3
Molecular Weight (g/mol) 302.41
Synonym 4-Hydroxyandrostendione, Formestane, Androst-4-ene-3,17-dione, 4-hydroxy-, 4-HAD, 4-Hydroxyandrost-4-ene-3,17-dione, 4-Hydroxyandrostene-3,17-dione, 4-Hydroxyandrostenedione, 4-OHA, CGP 32349, CRC 82/01, Formestane, Lentaron, NSC 282175
IUPAC Name (8R,9S,10R,13S,14S)-4-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-3,17-dione
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=C(O)C(=O)CC[C@]34C)[C@@H]1CCC2=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H26O3/c1-18-10-8-15(20)17(22)14(18)4-3-11-12-5-6-16(21)19(12,2)9-7-13(11)18/h11-13,22H,3-10H2,1-2H3/t11-,12-,13-,18+,19-/m0/s1
Formula Weight 302.1882
Percent Purity >95%
Chemical Name or Material Formestane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.