Learn More
Poulten Graf™ Fortuna™ Glass 1 Mark Bulb Pipette
Individually marked with the time of flow to transfer. Poulten Graf™ Fortuna™ Glass One Mark Bulb Pipettes are manufactured from clear soda lime glass and come with a batch certificate of conformity.
Brand:  Poulten Graf™ 1 2302502
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15922110
  • 22.75€ / Pack of 5
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Short practical delivery time
  • Tips and suction tubes reinforced
  • Acid and alkali resistant
Specifications
Specifications
AS
Yes
1 Pack
Bulb Pipet
Glass
Blue
2 mL