missing translation for 'onlineSavingsMsg'
Learn More

Fosaprepitant Dimeglumine, TRC

Product Code. 30136277
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136277 100 mg 100mg
30133529 25 mg 25mg
17932295 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136277 Supplier Toronto Research Chemicals Supplier No. TRCF727305100MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 265121-04-8
Molecular Formula C23 H22 F7 N4 O6 P . 2 C7 H17 N O5
Molecular Weight (g/mol) 1004.83
Synonym D-Glucitol, 1-deoxy-1-(methylamino)-, [3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-2,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]phosphonate (2:1), D-Glucitol, 1-deoxy-1-(methylamino)-, [3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-2,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]phosphonate (2:1) (salt) (9CI), Phosphonic acid, [3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-2,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]-, compd. with 1-deoxy-1-(methylamino)-D-glucitol (1:2) (9CI), Fosaprepitant di(N-methylglucamine), Fosaprepitant dimeglumine, MK 0517
IUPAC Name [3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-5-oxo-4H-1,2,4-triazol-1-yl]phosphonic acid;(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.C[C@@H](O[C@H]1OCCN(CC2=NN(C(=O)N2)P(=O)(O)O)[C@H]1c3ccc(F)cc3)c4cc(cc(c4)C(F)(F)F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H22F7N4O6P.2C7H17NO5/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)40-20-19(13-2-4-17(24)5-3-13)33(6-7-39-20)11-18-31-21(35)34(32-18)41(36,37)38;2*1-8-2-4(10)6(12)7(13)5(11)3-9/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H,31,32,35)(H2,36,37,38);2*4-13H,2-3H2,1H3/t12-,19+,20-;2*4-,5+,6+,7+/m100/s1
Formula Weight 1004.3379
Chemical Name or Material Fosaprepitant Dimeglumine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.