missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVP Antibody (VP2897R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08801APC
This item is not returnable.
View return policy
Description
MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MVP | |
| Monoclonal | |
| APC | |
| LRPVAULT1, Lung resistance-related protein, major vault protein | |
| Recombinant full-length human MVP protein (Uniprot: Q14764) | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 9961 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| VP2897R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction