missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxM1 Antibody (CL12131) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15129G
This item is not returnable.
View return policy
Description
FoxM1 Monoclonal antibody specifically detects FoxM1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| FoxM1 | |
| Monoclonal | |
| DyLight 488 | |
| FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN, Winged-helix factor from INS-1 cells | |
| This antibody was developed using a synthetic peptide derived from Q08050, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Mitotic Regulators | |
| 2305 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL12131 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction