missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR Antibody (CL13278) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18557C
This item is not returnable.
View return policy
Description
FoxO1/FKHR Monoclonal antibody specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot
Specifications
| FoxO1/FKHR | |
| Monoclonal | |
| DyLight 650 | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| This antibody was developed using a synthetic peptide derived from Q12778, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
| 2308 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL13278 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction