missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO3 Antibody (CL13603) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24565MFV450
This item is not returnable.
View return policy
Description
FOXO3 Monoclonal antibody specifically detects FOXO3 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| FOXO3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739 | |
| This antibody was developed using a synthetic peptide derived from O43524, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| 2309 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL13603 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction