missing translation for 'onlineSavingsMsg'
Learn More

FOXP4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen FOXP4
Concentration 800 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17649656
View Documents
Proteintech
29311-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17639656
View Documents
Proteintech
29311-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FOXP4 Polyclonal antibody apecifically detetects FOXP4 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

FOXP4
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Rabbit
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
116113
FOXP4 fusion protein Ag29953
Primary
Store at -20°C. Stable for one year after shipment.
800 μg/mL
Polyclonal
Liquid
Human, Mouse
BC052803
FOXP4
IgG
Antigen affinity purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.