Learn More
Invitrogen™ FPR2 Recombinant Rabbit Monoclonal Antibody (HL2664)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556716
This item is not returnable.
View return policy
Description
FPR2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Formyl peptide receptor 2 (FPR2) is a low affinity receptor for N-formyl-methionyl peptides. Binding of FMLP to FPR2 causes activation of neutrophils. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4.
Specifications
| FPR2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O88536, P25090 | |
| FPR2 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human FPRL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2664 | |
| Unconjugated | |
| FPR2 | |
| ALXR; E330010I07Rik; fMLPR1; FMLP-related receptor I; FMLP-R-I; FMLP-R-II; FMLPX; formyl peptide receptor 2; formyl peptide receptor, related sequence 2; formyl peptide receptor, related sequence 2-like; formyl peptide receptor-like 1; Formylpeptide receptor-related sequence 2; FPR2; FPR2A; FPRH1; FPRH2; Fprl1; Fprl-1; FPR-like 1; Fpr-rs1; Fpr-rs2; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); lipoxin A4 receptor-like protein; Lipoxin receptor; LXA4 receptor; LXA4R; N-formyl peptide receptor 2; N-formylpeptide receptor-like 2; RFP | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14289, 2358 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |