Learn More
FRA-1/FOSL1 Antibody (SR1652), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22276-100ul
This item is not returnable.
View return policy
Description
FRA-1/FOSL1 Monoclonal antibody specifically detects FRA-1/FOSL1 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| FRA-1/FOSL1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1652 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1 | |
| A synthesized peptide derived from human FRA-1/FOSL1 (Uniprot #: P15407) | |
| 100 μg | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 8061 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |