missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Antibody (FXN/2124), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79936F
This item is not returnable.
View return policy
Description
Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Frataxin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| FXN | |
| Recombinant fragment of human Frataxin (FXN) protein (around aa 57-210) (exact sequence is proprietary) (Uniprot: Q16595) | |
| 0.1 mL | |
| Diabetes Research | |
| Human | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FXN/2124 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction