Learn More
Hanna Instruments™ Free Chlorine Test Kit
Brand:  Hanna Instruments™ HI3831F
Additional Details : Weight : 0.03500kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 10115123
  • 46.00€ / Each
Estimated Shipment: 16-04-2024
to see stock.
Specifications
1 Each
0 to 2.5 mg/L
DPD
Free Chlorine Test Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 1 SET CHLORINE TEST KIT TOTAL
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric aicd, N,N-diethyl-1,4-phenylenediammonium sulphate, sodium hydroxide