Learn More
Hanna Instruments™ Free and Total Chlorine High Range Reagent
Product Code. 11703373
Change view
Click to view available options
For Use With (Application):
Photometric determination of free chlorine and total chlorine
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11703373 Photometric determination of free chlorine and total chlorine Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11703373 Supplier Hanna Instruments™ Supplier No. HI9373401
  • ONLINE EXCLUSIVE PRICE
    95.17€
    47.11€ / Pack of 100
    Save 48.06€
    50% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the photometric determination of free chlorine and total chlorine
  • Range : 0.00 to 10.0mg/L
  • Compatibility : HI-96734 portable photometer
Specifications
Type Free and Total Chlorine Reagent
For Use With (Application) Photometric determination of free chlorine and total chlorine