Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 5 in/127 mm
Description
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 5 in/127 mm, side access, stainless steel- Rack for 127 mm/5.0 in boxes (15/50 mL conical tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 681 (5.6 x 5.6 x 26.8 in)- Max. box number per rack: 5 (5 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 40 racks)- For ULT freezer: Innova C580 (max. 160 boxes), Innova C760 (max. 200 boxes), CryoCube™ FC660 series (max. 200 boxes)
Specifications
Specifications
| Product Type | Freezer Rack |