Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, MTP
Description
Freezer Rack: Innova™ C580, C760, locking rod, MTP, side access, stainless steel- Rack for MTP (18 mm)- Max. MTP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 140 x 91 x 766 (5.51 x 3.58 x 30.16)- Max. MTP number per rack: 38 (38 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 52 racks), Innova C760 (max. 66 racks)- For ULT freezer: Innova C580 (max. 1,976 MTP), Innova C760 (max. 2,508 MTP)- With locking rod for increased sample safety
Specifications
Specifications
| Product Type | Freezer Rack |