Learn More
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7
Description
Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F740 series (5-compartment, compartment 1-4), DWP (53 mm), side access, stainless steel- Rack for DWP- Max. DWP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 549 x 139 x 224 (21.6 x 5.5 x 8.8 in)- Max. DWP number per rack: 24 (4 x 6)- Stainless steel- For ULT freezer: Premium U410 (max. 15 racks), Premium U700 (max. 25 racks), HEF™ U410 (max. 15 racks), CryoCube™ F570 series (max. 25 racks), CryoCube F740 series (5-compartment version, compartment 1-4)- For ULT freezer: Premium U410 (max. 360 DWP), Premium U700 (max. 600 DWP), HEF™ U410 (max. 360 DWP), CryoCube™ F570 series (max. 600 DWP), CryoCube F740 series (5-compartment version, compartment 1-4)
Specifications
Specifications
| Product Type | Freezer Rack |