Learn More
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7
Product Code. 15713245
Product Code. 15713245 Supplier Eppendorf™ Supplier No. 6001021110
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F740 series (5-compartment, compartment 1-4), DWP (53 mm), side access, stainless steel- Rack for DWP- Max. DWP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 549 x 139 x 224 (21.6 x 5.5 x 8.8 in)- Max. DWP number per rack: 24 (4 x 6)- Stainless steel- For ULT freezer: Premium U410 (max. 15 racks), Premium U700 (max. 25 racks), HEF™ U410 (max. 15 racks), CryoCube™ F570 series (max. 25 racks), CryoCube F740 series (5-compartment version, compartment 1-4)- For ULT freezer: Premium U410 (max. 360 DWP), Premium U700 (max. 600 DWP), HEF™ U410 (max. 360 DWP), CryoCube™ F570 series (max. 600 DWP), CryoCube F740 series (5-compartment version, compartment 1-4)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack