Learn More
Brady™ FreezerBondz™ Ultra Thin Cryogenic Polyester Labels
Product Code. 11824205 Shop All Brady Products
Change view
Click to view available options
Quantity:
1500
2500/Roll
3000/Roll
5000/Roll
Dimensions (L x W):
12.70 x 25.4 mm (0.5 x 1 in.)
25.4 x 22.86 mm (1 x 0.9 in.)
25.4 x 48.26 mm (1 x 1.9 in.)
9.53 x 25.4 mm (0.37 x 1 in.)
Unit Size:
1 pack
1 roll
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensions (L x W) Unit Size
11824205 3000/Roll 25.4 x 48.26 mm (1 x 1.9 in.) 1 roll
10419473 5000/Roll 12.70 x 25.4 mm (0.5 x 1 in.) 1 pack
11814205 3000/Roll 25.4 x 22.86 mm (1 x 0.9 in.) 1 roll
11588834 2500/Roll 9.53 x 25.4 mm (0.37 x 1 in.) 1 roll
15148385 1500 25.4 x 48.26 mm (1 x 1.9 in.) 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11824205 Supplier Brady™ Supplier No. 805915
  • ONLINE EXCLUSIVE PRICE
    585.00€
    496.39€ / 1 rolls
    Save 88.61€
    15% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Durable Freezerbondz™ Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
  • Specialized — Aggressive adhesive B-492 FreezerBondz™ labels are ideal for curved, frozen surfaces like vials, centrifuge samples, tubes, and slides.
  • Durable — Labels withstand chemicals, abrasions, storage in liquid nitrogen, and a high/low temperature range of -320°F to 212°F (-196°C to 100°C).
  • Adhesive — Permanent acrylic adhesive for tight bonding onto smooth, curved surfaces, or wrapping around tubes and vials with a 1/8 in. overlap.
  • Certifications — RoHS compliant to 2011/65/EU (Pass 2015/863).
Spezifikation
Autoclavable Not Autoclavable
Certifications/Compliance RoHS compliant to 2011/65/EU
Description FreezerBondz Thermal Transfer Printer Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Polyester
Print Technology Thermal Transfer
Product Type Laboratory Label
Sterility Non-sterile
For Use With (Application) Laboratory identification, R6400 series ribbons, thermal transfer printers
For Use With (Equipment) BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Includes Roll of 3000 Labels
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
No. per Pack 3000 Label(s) / Roll
Quantity 3000/Roll
Shape Rectangle
Size 1 x 1.9 in. (L x W)
Size (Metric) 48.26 mm (W) x 25.40 mm (H)
Temperature Range (English) -320°F to 266°F
Temperature Range (Metric) -160°C to +130°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Vertical Repeat (Metric) 27.94 mm
Dimensions (L x W) 25.4 x 48.26 mm (1 x 1.9 in.)
Length (English) 1 in.
Width (English) 1.9 in.
Width (Metric) 48.26 mm
Height (English) 1 in.
Height (Metric) 25.40 mm
Alternate Part Number THT-156-492-3
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.