Learn More
Vileda Professional™ Cleanroom Squeegee with Swivel
Product Code. 17822486
Change view
Click to view available options
Length (English):
20 in.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
17822486 20 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17822486 Supplier Vileda Professional™ Supplier No. 119935
  • ONLINE EXCLUSIVE PRICE
    360.00€
    334.09€ / Pack of 10
    Save 25.91€
    7% Off
View all promotions
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Swivels 360 degrees allowing for full range of motion
  • Use on floors and walls after applying disinfectant
  • Squeegee spills to the drain in controlled environments
  • Can be autoclaved and exposed to gamma or EtO sterilization
  • Use with FHP Extendable Handle #119966
Specifications
Product Type Swivel Squeegee
For Use With (Application) Cleanroom applications
Length (English) 20 in.