missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRMD5 Antibody (CL9503) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88935V
This item is not returnable.
View return policy
Description
FRMD5 Monoclonal antibody specifically detects FRMD5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FRMD5 | |
| Monoclonal | |
| DyLight 405 | |
| FERM domain containing 5, FERM domain-containing protein 5, FLJ41022, MGC14161 | |
| This antibody was developed using a recombinant protein derived from Q7Z6J6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9503 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 84978 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction