Learn More
Invitrogen™ FSHR Recombinant Rabbit Monoclonal Antibody (HL2926)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556719
Product Code. 30275020
  • 500.00€ / 100µL
Description
FSHR Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Follicle-Stimulating Hormone Receptor (FSHR) expression has been reported in ovary, uterus, and testis. ESTs have been isolated from testis libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FSHR
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P23945, P35378
FSHR
Synthetic peptide encompassing a sequence within the Extracellular domain of human FSH Receptor. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL2926
Unconjugated
FSHR
follicle stimulating hormone receptor; follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; Fshr; FSH-R; FSHR1; FSHRO; LGR1; ODG1
Rabbit
Protein A
RUO
14309, 2492
-20°C, Avoid Freeze/Thaw Cycles