Learn More
FTCD Monoclonal Antibody (OTI8F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505005
Description
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.Specifications
| FTCD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95954 | |
| FTCD | |
| Full length human recombit protein of human FTCD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI8F12 | |
| Unconjugated | |
| FTCD | |
| LCHC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10841 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.