missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-60935AF488
This item is not returnable.
View return policy
Description
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.
Specifications
| FTO | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoblotting | |
| Q9C0B1 | |
| FTO | |
| A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Chicken, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoblot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 79068 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction