missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27707-1-AP-150UL
This item is not returnable.
View return policy
Description
Sodium-phosphate symporter which seems to play a fundamental housekeeping role in phosphate transport by absorbing phosphate from interstitial fluid for normal cellular functions such as cellular metabolism, signal transduction, and nucleic acid and lipid synthesis. In vitro, sodium-dependent phosphate uptake is not siginificantly affected by acidic and alkaline conditions, however sodium-independent phosphate uptake occurs at acidic conditions. May play a role in extracellular matrix, cartilage and vascular calcification. Functions as a retroviral receptor and confers human cells susceptibility to infection to amphotropic murine leukemia virus (A-MuLV), 10A1 murine leukemia virus (10A1 MLV) and some feline leukemia virus subgroup B (FeLV-B) variants.Specifications
| FTSJD2 | |
| Polyclonal | |
| Unconjugated | |
| CMTR1 | |
| CMTR1, FTSJD2, hMTr1, ISG95, KIAA0082, MTr1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23070, 74157 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N1G2, Q9DBC3 | |
| Cmtr1 | |
| FTSJD2 Fusion Protein Ag26722 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction