missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 8/FUT8 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00002530-B01P
This item is not returnable.
View return policy
Description
Fucosyltransferase 8/FUT8 Polyclonal antibody specifically detects Fucosyltransferase 8/FUT8 in Human samples. It is validated for Western Blot
Specifications
| Fucosyltransferase 8/FUT8 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Mouse | |
| IgG purified | |
| RUO | |
| 2530 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| alpha-(1,6)-fucosyltransferase, alpha1-6FucT, EC 2.4.1.68, Fucosyltransferase 8, fucosyltransferase 8 (alpha (1,6) fucosyltransferase), GDP-fucose--glycoprotein fucosyltransferase, GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase, Glycoprotein 6-alpha-L-fucosyltransferase, MGC26465 | |
| FUT8 (NP_835367.1, 1 a.a. - 575 a.a.) full-length human protein. MRPWTGSWRWIMLILFAWGTLLFYIGGHLVRDNDHPDHSSRELSKILAKLERLKQQNEDLRRMAESLRIPEGPIDQGPAIGRVRVLEEQLVKAKEQIENYKKQTRNGLGKDHEILRRRIENGAKELWFFLQSELKKLKNLEGNELQRHADEFLLDLGHHERSIMTDLYYLSQTDGAGDWREKEAKDLTELVQRRITYLQNPKDCSKAKKLVCNINKGCGYGCQLHHVVYCFMIAYGTQRTLILESQNWRYATGGWETVFRPVSETCTDRSGISTGHWSGEVKDKNVQVVELPIVDSLHPRPPYLPLAVPEDLADRLVRVHGDPAVWWVSQFVKYLIRPQPWLEKEIEEATKKLGFKHPVIGVHVRRTDKVGTEAAFHPIEEYMVHVEEHFQLLARRMQVDKKRVYLATDDPSLLKEAKTKYPNYEFISDNSISWSAGLHNRYTENSLRGVILDIHFLSQADFLVCTFSSQVCRVAYEIMQTLHPDASANFHSLDDIYYFGGQNAHNQIAIYAHQPRTADEIPMEPGDIIGVAGNHWDGYSKGVNRKLGRTGLYPSYKVREKIETVKYPTYPEAEK | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction