Learn More
Löser™ Funnel Cleaning Needle
Product Code. 11940557
Change view
Click to view available options
For Use With (Equipment):
Osmometer Type 15/16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11940557 Osmometer Type 15/16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11940557 Supplier Löser™ Supplier No. 0.01.0005
  • ONLINE EXCLUSIVE PRICE
    3.90€
    0.70€ / Pack of 1
    Save 3.20€
    82% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Funnel cleaning needle for i Osmometer
Funnel for cleaning the needle hole of automatic Osmometers
Specifications
Product Type Funnel Cleaning Needle
For Use With (Equipment) Osmometer Type 15/16
For Use With (Application) Cleaning needle and needle hole