missing translation for 'onlineSavingsMsg'
Learn More

Furafylline, TRC

Product Code. 30009250
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009250 5 mg 5mg
30006049 25 mg 25mg
30012570 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009250 Supplier Toronto Research Chemicals Supplier No. TRCF8640705MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80288-49-9
Molecular Formula C12 H12 N4 O3
Molecular Weight (g/mol) 260.25
Synonym 3-(2-furanylmethyl)-3,9-dihydro-1,8-dimethyl-1H-purine-2,6-dione
IUPAC Name 3-(furan-2-ylmethyl)-1,8-dimethyl-7H-purine-2,6-dione
SMILES CN1C(=O)N(Cc2occc2)c3nc(C)[nH]c3C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H12N4O3/c1-7-13-9-10(14-7)16(6-8-4-3-5-19-8)12(18)15(2)11(9)17/h3-5H,6H2,1-2H3,(H,13,14)
Formula Weight 260.0909
Percent Purity >95%
Chemical Name or Material Furafylline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.