missing translation for 'onlineSavingsMsg'
Learn More

Furamide, TRC

Product Code. 30400928
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400928 500 mg
30401861 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400928 Supplier Toronto Research Chemicals Supplier No. TRCF861000500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3736-81-0
Molecular Formula C14 H11 Cl2 N O4
Molecular Weight (g/mol) 328.15
Synonym 2-Furancarboxylic acid, 4-[(dichloroacetyl)methylamino]phenyl ester, 2-Furoic acid, ester with 2,2-dichloro-4'-hydroxy-N-methylacetanilide, Furoic acid, ester with 2,2-dichloro-4'-hydroxy-N-methylacetanilide, Acetanilide, 2,2-dichloro-4'-hydroxy-N-methyl-, 2-furoate (ester), 4-Hydroxy-N-methyl dichloroacetanilide 2-furoate, 8073 CB, Amebiazol, Dichlofurazol, Diclofurazol, Diloxanid furoate, Diloxanide furoate, Entamide 2-furoate, Entamide furoate, Furamide, Furamide (amebicide), Furentomin, Histomibal, Miforon
IUPAC Name [4-[(2,2-dichloroacetyl)-methylamino]phenyl] furan-2-carboxylate
SMILES CN(C(=O)C(Cl)Cl)c1ccc(OC(=O)c2occc2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H11Cl2NO4/c1-17(13(18)12(15)16)9-4-6-10(7-5-9)21-14(19)11-3-2-8-20-11/h2-8,12H,1H3
Formula Weight 327.0065
Chemical Name or Material Diloxanide Furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.