missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furfural-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Furfural-d4 |
| CAS | 1219803-80-1 |
| Molecular Formula | C5 2H4 O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]C(=O)c1oc([2 H])c([2 H])c1[2 H] |
| IUPAC Name | deuterio-(3,4,5-trideuteriofuran-2-yl)methanone |
| Molecular Weight (g/mol) | 100.11 |
| Formula Weight | 100.0462 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction