missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Mercaptan, TRC

Product Code. 30406873
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30406873 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30406873

Brand: Toronto Research Chemicals TRCF86430025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furan-2-ylmethanethiol
CAS 98-02-2
Molecular Formula C5 H6 O S
Quantity 25 g
Synonym 2-Furanmethanethiol (6CI, 7CI, 8CI, 9CI, ACI), (2-Furanyl)methylmercaptan, 2-(Mercaptomethyl)furan, 2-Furfurylmercaptan, 2-Furfurylthiol, 2-Furylmethanethiol, 2-Furylmethyl mercaptan, 2-Furylmethylthiol, Furan-2-ylmethanethiol, Furfuryl mercaptan, Furfuryl thiol, Furfurylthiol, NSC 41142
InChI Formula InChI=1S/C5H6OS/c7-4-5-2-1-3-6-5/h1-3,7H,4H2
SMILES SCc1occc1
IUPAC Name furan-2-ylmethanethiol
Molecular Weight (g/mol) 114.165
Formula Weight 114.014
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.