missing translation for 'onlineSavingsMsg'
Learn More

Furosemide Acyl-Beta-D-glucuronide, TRC

Product Code. 30011661
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011661 1 mg 1mg
30009412 10 mg 10mg
30007601 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011661 Supplier Toronto Research Chemicals Supplier No. TRCF8650101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72967-59-0
Molecular Formula C18 H19 Cl N2 O11 S
Molecular Weight (g/mol) 506.87
Synonym β-D-Glucopyranuronic acid, 1-[5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoate], Furosemide glucuronide, Furosemide Acyl-β-D-glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES NS(=O)(=O)c1cc(C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)c(NCc3occc3)cc1Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H19ClN2O11S/c19-9-5-10(21-6-7-2-1-3-30-7)8(4-11(9)33(20,28)29)17(27)32-18-14(24)12(22)13(23)15(31-18)16(25)26/h1-5,12-15,18,21-24H,6H2,(H,25,26)(H2,20,28,29)/t12-,13-,14+,15-,18-/m0/s1
Formula Weight 506.0398
Chemical Name or Material Furosemide Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.