missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUS Antibody (CL0190) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52874MFV500
This item is not returnable.
View return policy
Description
FUS Monoclonal antibody specifically detects FUS in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out,KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| FUS | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 75 kDa DNA-pairing protein, ALS6, amyotrophic lateral sclerosis 6, FUS1, fused in sarcoma, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, fusion, derived from t(12;16) malignant liposarcoma, fus-like protein, heterogeneous nuclear ribonucleoprotein P2, HNRNPP2, Oncogene FUS, Oncogene TLS, POMp75, RNA-binding protein FUS, TLShnRNP-P2, Translocated in liposarcoma protein | |
| This antibody was developed using a recombinant protein derived from P35637, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 2521 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0190 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction