Learn More
FXN Monoclonal Antibody (OTI3C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504335
Description
FXN is a mitochondrial protein which belongs to the FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia.Specifications
| FXN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16595 | |
| FXN | |
| Full length human recombit protein of human FXN produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3C7 | |
| Unconjugated | |
| FXN | |
| CyaY, FA, FARR, FRDA, X25 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2395 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.