missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97956-50ul
This item is not returnable.
View return policy
Description
FXR1 Polyclonal antibody specifically detects FXR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FXR1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| fragile X mental retardation syndrome-related protein 1, fragile X mental retardation, autosomal homolog 1, FXR1P, hFXR1p | |
| Produced in rabbits immunized with E. coli-derived Human FXR1 fragment. | |
| 50 μg | |
| Apoptosis, Immune System Diseases, Immunology | |
| 8087 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction