missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83432
This item is not returnable.
View return policy
Description
FXR2 Polyclonal antibody specifically detects FXR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| FXR2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ERLQIDEQLRQIGLGFRPPGSGRGSGGSDKAGYSTDESSSSSLHATRTYGGSYGGRGRGRRTGGPAYGPSSDVSTAS | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience | |
| 9513 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction