Learn More
FXYD5/Dysadherin Antibody (CL14037) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24585PCP
Product Code. 30298822
  • 737.00€ / 0.10mL
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FXYD5/Dysadherin
Monoclonal
PerCP
DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC
This antibody was developed using a recombinant protein derived from Q96DB9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
53827
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL14037