Learn More
Bacharach™ Fyrite™ CO2 or O2 Analyzer Refill Kits
Product Code. 11759778
Encompass_Preferred
Change view
Click to view available options
Includes:
3 Bottles of fluid (7.6% CO2)
Concentration Measurement Range:
0 to 7.6% CO
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Concentration Measurement Range unitSize
11759778 3 Bottles of fluid (7.6% CO2) 0 to 7.6% CO Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759778 Supplier Bacharach™ Supplier No. 105100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Keep your analyzer working with the Fyrite CO2 or O2 Analyzer Refill Kit.
  • Include two bottles of fluid, top gasket screws, filtering material
  • 10-884-6 consists of three bottles of fluid only
Specifications
Concentration Measurement Range 0 to 7.6% CO
Type CO2 Analyzer Refill Kit
Includes 3 Bottles of fluid (7.6% CO2)
compliance-icons compliance-icons
Product Identifier
  • fluid fyrite CO2 7%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands, forearms, andother exposed areas thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see section 4 on this label).
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide,alcohols C7-C9, C.I. acid red 14, 2-naphthalenol