missing translation for 'onlineSavingsMsg'
Learn More

Syngene G:BOX - F9 with Feedback Lens
GREENER_CHOICE

Product Code. 31749210 Shop All Syngene Products
Change view
Click to view available options
Pixel Size:
9 MP
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pixel Size unitSize
31749210 9 MP 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749210 Supplier Syngene Supplier No. F9LFB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

G:BOX with Feedback Lens allows for automated imaging of DNA gels and visible protein gels.

A gel documentation system for large gels capable of basic imaging applications including DNA and Protein gels.

  • Camera capable of producing 16 bit images with 65535 grey scales.
  • F1.2 motorized zoom lens with feedback.
  • Comes with a seven position filter wheel and epi white light.
  • Option to upgrade the system for other applications in the future.
  • Maximum viewing area available is 32.5 x 24.1 cm
  • PC required (not included).

GeneSys is an application driven software designed for rapid image capture of biological samples. It has a simple icon based user interface usable on Windows 10, 8 and 7 platforms.

  • Takes the guess work out of imaging, and produces quick and simple publication quality images.
  • All files produced are a unique SGD format, where only raw data is analysed for quantification of samples.
  • Files can also be saved as TIFF, JPEG, BMP formats.
  • User protocols can be saved to review previous images captured, and easily replicate future imaged gels.
  • All captured images can be simply edited with annotations, edit tools, image sharpen and exported to GeneTools.

GeneTools software provides the user with rapid quantification of samples and goes from blots to analysed data in a few quick steps. Analysis sample types include:

  • Molecular weight
  • Colony counting
  • Spot blot
  • Band matching

All samples can be quantified, including multiplexed protein fluorescence analysis of up to 5 samples at once. Raw data files can be quickly quantified to produce true data for publication data. High quality reports can be exported in Word and Excel files for simple review. Both GeneSys and GeneTools software meet the requirements for 21 CFR Part 11 and GLP environments. The system comes with a 3 year warranty (return to factory), unlimited licences of GeneTools analysis software and twice annual free software updates. IQ/OQ is available upon request.

Specifications

Voltage 230 V
Depth (English) 17.7 in.
Depth (Metric) 45 cm
Filter Type 7-position Filter Wheel
Height (English) 33.1 in.
Filter Area Up to 25 x 30 cm
Pixel Size 9 MP
Width (English) 22.4 in.
Height (Metric) 84 cm
Operating System Windows 7 or above
Software GeneSys and GeneTools Included (Unlimited)
Width (Metric) 57 cm
Lens Type Motorized zoom with Feedback
Hood Type Removable
Size 57 x 84 x 45 cm
For Use With (Application) Visible Blots
Product Type Gel Documentation System
Display Type None
Frequency 50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.