Learn More
IKA™ G-L Tube Adapter
Product Code. 16288701 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
1.5 to 2 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16288701 1.5 to 2 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16288701 Supplier IKA™ Supplier No. 0030001347
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
G-L Tube Adapter for use with 1.5 to 2 mL rotor.
  • Adapter
  • 0.5 mL Tube
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1.5 to 2 mL
Description Diameter: 8 mm
Includes 6/Pk