Learn More
G3BP2 Mouse, anti-Human, Clone: 2E5G3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68580-1-IG-150UL
 View more versions of this product
Product Code. 18006215
  • 410.00€ / 150µL
Description
The G3BP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G3BP2. This antibody recognizes human, rat antigen. The G3BP2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
G3BP2
2E5G3
G3BP 2, G3BP2, KIAA0660
150 μL
9908
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunofluorescence, ELISA
1000 μg/mL
Mouse
Primary
Human, Rat
150 kDa