missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | G5pr |
| Synonym | C14orf10, chromosome 14 open reading frame 10, FLJ20644, G4-1, G5pr, protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma, protein phosphatase 2, regulatory subunit B'', gamma, Protein phosphatase subunit G5PR, rhabdomyosarcoma antigen MU-RMS-40.6A, Rhabdomyosarcoma antigen MU-RMS-40.6A/6C, rhabdomyosarcoma antigen Mu-RMS-40.6C, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma |
| Gene Symbol | PPP2R3C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?