missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6PC Antibody (CL5817) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61432AF405
This item is not returnable.
View return policy
Description
G6PC Monoclonal antibody specifically detects G6PC in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| G6PC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 | |
| This antibody was developed using a synthetic peptide derived from P35575, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5817 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2538 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction