missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-199-0.02ML
This item is not returnable.
View return policy
Description
GABA-A R beta 3 Polyclonal specifically detects GABA-A R beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
| GABA-A R beta 3 | |
| Polyclonal | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:300, Immunohistochemistry Free-Floating, Immunofluorescence 1:300 | |
| P63079 | |
| GABRB3 | |
| Fusion protein from the cytoplasmic loop of the beta 3 subunit | |
| Affinity Purified | |
| RUO | |
| 2562 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051 | |
| Rabbit | |
| 53 kDa | |
| 0.02 mL | |
| Primary | |
| Specific for the ∼53k beta 3-subunit of the GABAA receptor in Western blots. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction