Learn More
Gabapentin Related Compound A, TRC
Product Code. 30407741
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407741 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30407741 Supplier Toronto Research Chemicals Supplier No. TRCG117275250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,3-Pentamethylene-4-butyrolactam
CAS 64744-50-9
Purity Grade Notes HPLC
Molecular Formula C9 H15 N O
Quantity 250 mg
Synonym 2-Azaspiro[4.5]decan-3-one, Gabapentin Imp. A (EP), 3,3-Pentamethylene-4-butyrolactam
InChI Formula InChI=1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11)
SMILES O=C1CC2(CCCCC2)CN1
IUPAC Name 2-azaspiro[4.5]decan-3-one
Molecular Weight (g/mol) 153.22
Formula Weight 153.1154
Percent Purity >95