missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD2/GAD65 Antibody (CL2736) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46629UV
This item is not returnable.
View return policy
Description
GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GAD2/GAD65 | |
| Monoclonal | |
| DyLight 350 | |
| EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
| This antibody was developed using a recombinant protein derived from Q05329, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
| 2572 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2736 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction