missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD45 alpha Antibody (OTI1C9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70559B
This item is not returnable.
View return policy
Description
GADD45 alpha Monoclonal specifically detects GADD45 alpha in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GADD45 alpha | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GADD45A | |
| Full length recombinant protein of human GADD45 alpha (NP_001915) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| OTI1C9 | |
| Western Blot | |
| DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 1647.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction